Assay Details
Target Gene Details
Entrez Gene ID: | 10602 |
Gene Name: | CDC42 effector protein 3 |
Gene Aliases: |
BORG2, CEP3, UB1 |
Location: |
Chr.2:37641882-37672535 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC42EP3 | NM_001270436.1 | NP_001257365.1 | ||
| NM_001270437.1 | NP_001257366.1 | |||
| NM_001270438.1 | NP_001257367.1 | |||
| NM_006449.4 | NP_006440.2 | |||
| AK128735.1 | ||||
| AK313483.1 | ||||
| AL136842.1 | CAB66776.1 | |||
| AL538577.3 | ||||
| BC019270.1 | AAH19270.1 | |||
| BQ067953.1 | ||||
| DB121680.1 | ||||
| HY003915.1 | ||||
| HY017509.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv998567 | Chr.2:37659101 - 37683484 on Build GRCh38 | Gain |
|
| nsv833825 | Chr.2:37581026 - 37758774 on Build GRCh38 | Gain |
|
| esv2759043 | Chr.2:37569511 - 37911796 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map