Assay Details
Target Gene Details
Entrez Gene ID: | 79843 |
Gene Name: | family with sequence similarity 124 member B |
Gene Aliases: |
- |
Location: |
Chr.2:224378698-224401994 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM124B | NM_001122779.1 | NP_001116251.1 | ||
| NM_024785.2 | NP_079061.2 | |||
| AK026399.1 | BAB15471.1 | |||
| AK075126.1 | ||||
| BC025754.1 | AAH25754.1 |
More Information
Set Membership: |
|

Genomic Map