Assay Details
Target Gene Details
Entrez Gene ID: | 51594 |
Gene Name: | neuroblastoma amplified sequence |
Gene Aliases: |
ILFS2, NAG, SOPH |
Location: |
Chr.2:14998067-15561348 on Build GRCh38 |
Assay Gene Location: | Within Intron 54 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NBAS | XM_017004317.1 | XP_016859806.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv515567 | Chr.2:14864624 - 15766654 on Build GRCh38 | Gain+Loss |
|
| esv3589906 | Chr.2:15024706 - 15348523 on Build GRCh38 | Loss |
|
| esv3589903 | Chr.2:15016319 - 15057663 on Build GRCh38 | Loss |
|
| esv3589904 | Chr.2:15016426 - 15121840 on Build GRCh38 | Loss |
|
| esv3589902 | Chr.2:15003909 - 15076710 on Build GRCh38 | Loss |
|
| esv3891570 | Chr.2:15019496 - 15054435 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map