Assay Details
Target Gene Details
Entrez Gene ID: | 89782 |
Gene Name: | leishmanolysin like peptidase |
Gene Aliases: |
GP63, INV, IX14, MSP |
Location: |
Chr.3:197960200-198043720 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMLN | NM_001136049.2 | NP_001129521.2 | ||
| NM_033029.3 | NP_149018.2 | |||
| NR_026786.1 | ||||
| NR_026787.1 | ||||
| XM_011513270.1 | XP_011511572.1 | |||
| XM_011513271.1 | XP_011511573.1 | |||
| XM_017007428.1 | XP_016862917.1 | |||
| AJ312398.1 | CAC42882.1 | |||
| AJ312399.1 | CAC42883.1 | |||
| AK299119.1 | ||||
| AL832783.1 | ||||
| AM920777.1 | ||||
| AM920778.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3599331 | Chr.3:197953544 - 197984148 on Build GRCh38 | Gain |
|
| esv2763718 | Chr.3:197746211 - 198169247 on Build GRCh38 | Gain+Loss |
|
| dgv5045n100 | Chr.3:197843263 - 198129562 on Build GRCh38 | Gain |
|
| nsv1013844 | Chr.3:197902008 - 197975418 on Build GRCh38 | Gain |
|
| nsv527527 | Chr.3:197842688 - 198087104 on Build GRCh38 | Gain |
|
| esv2751989 | Chr.3:197748569 - 198106887 on Build GRCh38 | Gain |
|
| esv3893855 | Chr.3:197912510 - 198111391 on Build GRCh38 | Gain |
|
| nsv1009299 | Chr.3:197916770 - 198018555 on Build GRCh38 | Gain |
|
| nsv999711 | Chr.3:197871402 - 198011040 on Build GRCh38 | Gain |
|
| esv3599324 | Chr.3:197837661 - 197996085 on Build GRCh38 | Gain |
|
| dgv5044n100 | Chr.3:197821762 - 197975418 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77259890] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map