Assay Details
Target Gene Details
Entrez Gene ID: | 285386 |
Gene Name: | tumor protein p63 regulated 1 |
Gene Aliases: |
FAM79B |
Location: |
Chr.3:189000774-189325304 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPRG1 | NM_198485.3 | NP_940887.1 | ||
| XM_005247382.4 | XP_005247439.1 | |||
| XM_011512732.2 | XP_011511034.2 | |||
| XM_011512734.2 | XP_011511036.1 | |||
| XM_011512735.2 | XP_011511037.1 | |||
| XM_017006257.1 | XP_016861746.1 | |||
| XM_017006258.1 | XP_016861747.1 | |||
| XM_017006259.1 | XP_016861748.1 | |||
| XM_017006260.1 | XP_016861749.1 | |||
| XM_017006261.1 | XP_016861750.1 | |||
| XM_017006262.1 | XP_016861751.1 | |||
| XM_017006263.1 | XP_016861752.1 | |||
| AK123232.1 | BAC85565.1 | |||
| AK125682.1 | BAC86245.1 | |||
| BC101550.1 | AAI01551.1 | |||
| BC101552.1 | AAI01553.1 |
Target Gene Details
Entrez Gene ID: | 100874027 |
Gene Name: | TPRG1 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.3:189238686-189240594 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPRG1-AS2 | NR_046722.1 | |||
| BG213396.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829819 | Chr.3:189126667 - 189280619 on Build GRCh38 | Gain |
|

Genomic Map