Assay Details
Target Gene Details
Entrez Gene ID: | 131118 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C19 |
Gene Aliases: |
PAM18, TIM14, TIMM14 |
Location: |
Chr.3:180983709-180989774 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC19 | NM_001190233.1 | NP_001177162.1 | ||
| NM_145261.3 | NP_660304.1 | |||
| NR_033721.1 | ||||
| NR_033722.1 | ||||
| NR_033723.1 | ||||
| NR_046073.1 | ||||
| AA775489.1 | ||||
| AI268262.1 | ||||
| AK307097.1 | ||||
| AK309019.1 | ||||
| AK311765.1 | ||||
| AW161751.1 | ||||
| BC009702.1 | AAH09702.1 | |||
| BC073989.1 | AAH73989.1 | |||
| BE773017.1 | ||||
| BF666599.1 | ||||
| BG492744.1 | ||||
| BM701756.1 | ||||
| BM702203.1 | ||||
| BQ438379.1 | ||||
| CB052107.1 | ||||
| CB159371.1 | ||||
| DA158066.1 |
More Information
Set Membership: |
|

Genomic Map