Assay Details
Target Gene Details
Entrez Gene ID: | 101927832 |
Gene Name: | PAQR9 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:142964058-143001090 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAQR9-AS1 | NR_125393.1 | |||
| DB456045.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs200173755,rs202025608] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map