Assay Details
Target Gene Details
Entrez Gene ID: | 54625 |
Gene Name: | poly(ADP-ribose) polymerase family member 14 |
Gene Aliases: |
ARTD8, BAL2, PARP-14, pART8 |
Location: |
Chr.3:122680726-122730840 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARP14 | NM_017554.2 | NP_060024.2 | ||
| XM_011512928.2 | 1 | 104 | XP_011511230.1 | |
| XM_011512929.2 | 1 | 112 | XP_011511231.1 | |
| AK304269.1 | ||||
| CR977108.1 | ||||
| DC320673.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829705 | Chr.3:122595197 - 122741131 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map