Assay Details
Target Gene Details
Entrez Gene ID: | 132204 |
Gene Name: | synaptoporin |
Gene Aliases: |
SPO |
Location: |
Chr.3:63200604-63616924 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYNPR | XM_017005731.1 | XP_016861220.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv76n68 | Chr.3:63019671 - 63243247 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114659841,rs72876365] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map