Assay Details
Target Gene Details
Entrez Gene ID: | 2912 |
Gene Name: | glutamate metabotropic receptor 2 |
Gene Aliases: |
GLUR2, GPRC1B, MGLUR2, mGlu2 |
Location: |
Chr.3:51707033-51718616 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508921 | Chr.3:51647199 - 51803073 on Build GRCh38 | Insertion |
|
| nsv954856 | Chr.3:51687685 - 51721184 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map