Assay Details
Target Gene Details
Entrez Gene ID: | 26355 |
Gene Name: | family with sequence similarity 162 member A |
Gene Aliases: |
C3orf28, E2IG5, HGTD-P |
Location: |
Chr.3:122384176-122410114 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM162A | NM_014367.3 | NP_055182.3 | ||
| AF107495.1 | AAG39275.1 | |||
| AF191020.1 | AAF09484.1 | |||
| AF201944.1 | AAF86880.1 | |||
| AF250321.1 | AAG44569.1 | |||
| AK026030.1 | ||||
| AK293994.1 | ||||
| BC010896.1 | AAH10896.1 | |||
| BC015060.1 | AAH15060.1 | |||
| BI460077.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3973 | Chr.3:122372209 - 122416890 on Build GRCh38 | Deletion |
|
| esv3597609 | Chr.3:122393677 - 122407495 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map