Assay Details
Target Gene Details
Entrez Gene ID: | 347736 |
Gene Name: | NME/NM23 family member 9 |
Gene Aliases: |
NM23-H9, TXL-2, TXL2, TXNDC6 |
Location: |
Chr.3:138261437-138331785 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 13 - Exon 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428422 | Chr.3:137344347 - 138686784 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map