Assay Details
Target Gene Details
Entrez Gene ID: | 205428 |
Gene Name: | chromosome 3 open reading frame 58 |
Gene Aliases: |
DIA1, GoPro49, HASF |
Location: |
Chr.3:143971798-143992368 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C3orf58 | NM_001134470.1 | NP_001127942.1 | ||
| NM_173552.3 | NP_775823.1 | |||
| XM_011512543.2 | XP_011510845.1 | |||
| XM_017005869.1 | XP_016861358.1 | |||
| AK095161.1 | ||||
| AK095400.1 | ||||
| AK294020.1 | ||||
| AK315082.1 | ||||
| AL832839.1 | ||||
| BC037293.2 | AAH37293.1 | |||
| DC310234.1 | ||||
| DC310421.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893777 | Chr.3:143882155 - 144016702 on Build GRCh38 | Gain |
|
| esv2726004 | Chr.3:143978274 - 143979033 on Build GRCh38 | Deletion |
|
| esv2726007 | Chr.3:143978596 - 143978748 on Build GRCh38 | Deletion |
|
| nsv518814 | Chr.3:143947983 - 143984908 on Build GRCh38 | Gain |
|
| nsv591922 | Chr.3:143926400 - 144106597 on Build GRCh38 | Loss |
|
| esv2321998 | Chr.3:143978428 - 143978855 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76723797] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map