Assay Details
Target Gene Details
Entrez Gene ID: | 100507291 |
Gene Name: | uncharacterized LOC100507291 |
Gene Aliases: |
- |
Location: |
Chr.3:139389803-139583319 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100507291 | NR_121609.1 | |||
| DA364121.1 |
Target Gene Details
Entrez Gene ID: | 5947 |
Gene Name: | retinol binding protein 1 |
Gene Aliases: |
CRABP-I, CRBP, CRBP1, CRBPI, RBPC |
Location: |
Chr.3:139517434-139539829 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBP1 | NM_001130992.1 | NP_001124464.1 | ||
| NM_001130993.1 | NP_001124465.1 | |||
| NM_002899.3 | NP_002890.2 | |||
| AK290315.1 | ||||
| AK301684.1 | ||||
| AK307378.1 | ||||
| AK309492.1 | ||||
| BC121052.2 | ||||
| BU734286.1 | ||||
| CR541979.1 | CAG46776.1 | |||
| CR542005.1 | CAG46802.1 | |||
| DC394326.1 | ||||
| M11433.1 | AAA60257.1 | |||
| M36809.1 | AAA35714.1 |
More Information
Set Membership: |
|

Genomic Map