Assay Details
Target Gene Details
Entrez Gene ID: | 59345 |
Gene Name: | G protein subunit beta 4 |
Gene Aliases: |
CMTD1F |
Location: |
Chr.3:179396088-179451583 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNB4 | NM_021629.3 | 10 | 4392 | NP_067642.1 |
| XM_006713721.2 | 10 | 4323 | XP_006713784.1 | |
| AK091332.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4139 | Chr.3:179384796 - 179418003 on Build GRCh38 | Insertion |
|
| nsv829806 | Chr.3:179234575 - 179436585 on Build GRCh38 | Gain |
|
| dgv4999n100 | Chr.3:179227018 - 179528391 on Build GRCh38 | Gain |
|
| nsv592650 | Chr.3:179250846 - 179534475 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map