Assay Details
Target Gene Details
Entrez Gene ID: | 100506840 |
Gene Name: | IQ motif containing F5 pseudogene |
Gene Aliases: |
- |
Location: |
Chr.3:51817603-51819635 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IQCF4 | NR_038213.1 | |||
| NR_038214.1 | ||||
| NR_038215.1 | ||||
| AA398133.1 | ||||
| AA460742.1 | ||||
| GQ870251.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1139789 | Chr.3:51816784 - 51820284 on Build GRCh38 | Duplication |
|
| nsv520211 | Chr.3:51738331 - 52132131 on Build GRCh38 | Gain |
|

Genomic Map