Assay Details
Target Gene Details
Entrez Gene ID: | 60675 |
Gene Name: | prokineticin 2 |
Gene Aliases: |
BV8, HH4, KAL4, MIT1, PK2 |
Location: |
Chr.3:71771655-71785206 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PROK2 | NM_001126128.1 | NP_001119600.1 | ||
| NM_021935.3 | NP_068754.1 | |||
| XM_017006974.1 | XP_016862463.1 | |||
| AF182069.2 | AAG16893.2 | |||
| AF333025.1 | AAK49919.1 | |||
| AY349131.1 | AAR06657.1 | |||
| BC069395.1 | AAH69395.1 | |||
| BC096695.1 | AAH96695.1 | |||
| BC098110.1 | AAH98110.1 | |||
| BC098162.1 | AAH98162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv590600 | Chr.3:71717301 - 71780868 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map