Assay Details
Target Gene Details
Entrez Gene ID: | 101927597 |
Gene Name: | NCK1 antisense RNA 1 (head to head) |
Gene Aliases: |
SLC35G2-AS1 |
Location: |
Chr.3:136841976-136862063 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCK1-AS1 | NR_110175.1 | 1 | 81 | |
| NR_110176.1 | 1 | 81 | ||
| NR_110177.1 | 1 | 81 | ||
| CB047433.1 | 1 | 81 | ||
| DA714434.1 | ||||
| HY007950.1 | 1 | 81 |

Genomic Map