Assay Details
Target Gene Details
Entrez Gene ID: | 55167 |
Gene Name: | male-specific lethal 2 homolog (Drosophila) |
Gene Aliases: |
MSL-2, MSL2L1, RNF184 |
Location: |
Chr.3:136148918-136197300 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MSL2 | NM_001145417.1 | NP_001138889.1 | ||
| NM_018133.3 | NP_060603.2 | |||
| XM_006713684.3 | XP_006713747.1 | |||
| XM_011512949.2 | XP_011511251.1 | |||
| AB046805.1 | ||||
| AK091956.1 | ||||
| AK292557.1 | ||||
| AK302491.1 | ||||
| AL834289.1 | CAD38963.2 | |||
| BC093764.1 | AAH93764.1 | |||
| BC093790.1 | AAH93790.1 | |||
| DC409643.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv955351 | Chr.3:136163759 - 136195858 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115068496] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map