Assay Details
Target Gene Details
Entrez Gene ID: | 165631 |
Gene Name: | poly(ADP-ribose) polymerase family member 15 |
Gene Aliases: |
ARTD7, BAL3, pART7 |
Location: |
Chr.3:122575914-122639047 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARP15 | XM_005247159.3 | 1 | 1358 | XP_005247216.1 |
| XM_005247160.3 | 1 | 1358 | XP_005247217.1 | |
| XM_011512475.2 | 1 | 1362 | XP_011510777.1 | |
| XM_011512476.2 | 1 | 1361 | XP_011510778.1 | |
| XM_011512477.2 | 1 | 1358 | XP_011510779.1 | |
| XM_011512480.2 | 1 | 1374 | XP_011510782.1 | |
| XM_017005791.1 | 1 | 1358 | XP_016861280.1 |
More Information
Set Membership: |
|

Genomic Map