Assay Details
Target Gene Details
Entrez Gene ID: | 101927111 |
Gene Name: | SUCLG2 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.3:67654697-67947713 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUCLG2-AS1 | NR_109992.1 | |||
| NR_109993.1 | ||||
| AL832184.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003158 | Chr.3:66999779 - 68395539 on Build GRCh38 | Loss |
|
| nsv470638 | Chr.3:67316917 - 67764314 on Build GRCh38 | Loss |
|

Genomic Map