Assay Details
Target Gene Details
Entrez Gene ID: | 64770 |
Gene Name: | coiled-coil domain containing 14 |
Gene Aliases: |
- |
Location: |
Chr.3:123887383-123961412 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC14 | XM_005247715.4 | XP_005247772.1 | ||
| XM_011513081.2 | XP_011511383.1 | |||
| XM_017007061.1 | XP_016862550.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3597633 | Chr.3:123813223 - 123952334 on Build GRCh38 | Gain |
|
| nsv591428 | Chr.3:123608828 - 124139902 on Build GRCh38 | Gain |
|
| nsv3979 | Chr.3:123854567 - 123898685 on Build GRCh38 | Insertion |
|
| nsv1007525 | Chr.3:123876493 - 123958230 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map