Assay Details
Target Gene Details
Entrez Gene ID: | 5096 |
Gene Name: | propionyl-CoA carboxylase beta subunit |
Gene Aliases: |
- |
Location: |
Chr.3:136250325-136330171 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCCB | NM_000532.4 | NP_000523.2 | ||
| NM_001178014.1 | NP_001171485.1 | |||
| XM_011512873.1 | XP_011511175.1 | |||
| AB209009.1 | BAD92246.1 | |||
| AF217984.1 | AAG17227.1 | |||
| AK130359.1 | ||||
| AK225215.1 | ||||
| AK225733.1 | ||||
| AK295312.1 | ||||
| AK302522.1 | ||||
| AK303079.1 | ||||
| AL831978.2 | CAD89898.1 | |||
| BC013768.1 | AAH13768.1 | |||
| BC053661.1 | AAH53661.1 | |||
| S67325.1 | AAB28900.1 | |||
| X73424.1 | CAA51825.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508246 | Chr.3:136203181 - 136319780 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map