Assay Details
Target Gene Details
Entrez Gene ID: | 57092 |
Gene Name: | PEST proteolytic signal containing nuclear protein |
Gene Aliases: |
- |
Location: |
Chr.3:101574094-101594465 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
More Information
Set Membership: |
|

Genomic Map