Assay Details
Target Gene Details
Entrez Gene ID: | 101928376 |
Gene Name: | IL12A antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:159913400-160207092 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL12A-AS1 | NR_108088.1 | |||
| AK097161.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997776 | Chr.3:159867308 - 159950633 on Build GRCh38 | Gain |
|
| nsv4087 | Chr.3:159896535 - 159929774 on Build GRCh38 | Insertion |
|
| esv3584505 | Chr.3:159889156 - 159930826 on Build GRCh38 | Gain |
|
| nsv829769 | Chr.3:159889124 - 160087495 on Build GRCh38 | Gain |
|
| nsv4086 | Chr.3:159894732 - 159939992 on Build GRCh38 | Deletion |
|

Genomic Map