Assay Details
Target Gene Details
Entrez Gene ID: | 100874043 |
Gene Name: | TPRG1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:188941715-188947639 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPRG1-AS1 | NR_046873.1 | |||
| BG778254.1 |

Genomic Map