Assay Details
Target Gene Details
Entrez Gene ID: | 5580 |
Gene Name: | protein kinase C delta |
Gene Aliases: |
ALPS3, CVID9, MAY1, PKCD, nPKC-delta |
Location: |
Chr.3:53161207-53192717 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKCD | NM_001316327.1 | NP_001303256.1 | ||
| NM_006254.3 | NP_006245.2 | |||
| NM_212539.1 | NP_997704.1 | |||
| XM_006713259.2 | XP_006713322.1 | |||
| XM_017006856.1 | XP_016862345.1 | |||
| AK313216.1 | ||||
| BC043350.1 | AAH43350.1 | |||
| L07860.1 | AAA03176.1 | |||
| L07861.1 | AAA03175.1 | |||
| Z22521.1 | CAA80249.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv460554 | Chr.3:53145738 - 53194903 on Build GRCh38 | Loss |
|
| nsv954863 | Chr.3:53148285 - 53166784 on Build GRCh38 | Deletion |
|
| nsv834699 | Chr.3:53117860 - 53291778 on Build GRCh38 | Loss |
|
| nsv517233 | Chr.3:53141541 - 53286984 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map