Assay Details
Target Gene Details
Entrez Gene ID: | 151835 |
Gene Name: | copine family member 9 |
Gene Aliases: |
- |
Location: |
Chr.3:9703807-9729908 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPNE9 | NM_153635.2 | NP_705899.2 | ||
| XM_011533386.2 | XP_011531688.1 | |||
| XM_017005747.1 | XP_016861236.1 | |||
| AA758411.1 | ||||
| BC130375.1 | ||||
| BU101942.1 | ||||
| CA307982.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010768 | Chr.3:9502870 - 9873902 on Build GRCh38 | Gain |
|
| nsv1008805 | Chr.3:9694224 - 9756456 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map