Assay Details
Target Gene Details
Entrez Gene ID: | 6478 |
Gene Name: | siah E3 ubiquitin protein ligase 2 |
Gene Aliases: |
hSiah2 |
Location: |
Chr.3:150741123-150763476 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIAH2 | NM_005067.5 | NP_005058.3 | ||
| AK223565.1 | BAD97285.1 | |||
| BC013082.2 | AAH13082.1 | |||
| U76248.1 | AAC51908.1 | |||
| Y15268.1 | CAA75557.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73010939] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map