Assay Details
Target Gene Details
Entrez Gene ID: | 152273 |
Gene Name: | FYVE, RhoGEF and PH domain containing 5 |
Gene Aliases: |
ZFYVE23 |
Location: |
Chr.3:14818962-14934565 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGD5 | NM_001320276.1 | NP_001307205.1 | ||
| NM_152536.3 | NP_689749.3 | |||
| XM_011533422.1 | XP_011531724.1 | |||
| XM_011533423.1 | XP_011531725.1 | |||
| AK097276.1 | BAC04989.1 | |||
| AK131078.1 | BAC85128.1 | |||
| BC132814.1 | ||||
| BC132816.1 | ||||
| BC144300.1 | ||||
| BX538312.1 | CAD98090.1 | |||
| BX640820.1 | CAE45896.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893664 | Chr.3:14808160 - 14845522 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73814195] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map