Assay Details
Target Gene Details
Entrez Gene ID: | 100507291 |
Gene Name: | uncharacterized LOC100507291 |
Gene Aliases: |
- |
Location: |
Chr.3:139389803-139583319 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100507291 | NR_121608.1 | |||
| NR_121609.1 | ||||
| BX098847.1 | ||||
| DA364121.1 | ||||
| DB034152.1 | ||||
| DB446595.1 |

Genomic Map