Assay Details
Target Gene Details
Entrez Gene ID: | 9175 |
Gene Name: | mitogen-activated protein kinase kinase kinase 13 |
Gene Aliases: |
LZK, MEKK13, MLK |
Location: |
Chr.3:185282938-185489094 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K13 | NM_001242314.1 | NP_001229243.1 | ||
| NM_001242317.1 | NP_001229246.1 | |||
| NR_038322.1 | ||||
| XM_017007456.1 | XP_016862945.1 | |||
| XM_017007458.1 | XP_016862947.1 | |||
| AK297646.1 | ||||
| AK312714.1 | ||||
| BQ432918.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829817 | Chr.3:185227125 - 185409387 on Build GRCh38 | Gain |
|
| esv3598966 | Chr.3:185273685 - 185354155 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map