Assay Details
Target Gene Details
Entrez Gene ID: | 100132677 |
Gene Name: | BSN antisense RNA 2 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.3:49549306-49554366 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BSN-AS2 | NR_038866.1 | 3 | 1565 | |
| AK096387.1 | 3 | 1565 | BAC04774.1 | |
| AK097709.1 | 4 | 1407 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3369484 | Chr.3:49550015 - 49552013 on Build GRCh38 | Duplication |
|
| nsv834687 | Chr.3:49411199 - 49594355 on Build GRCh38 | Loss |
|

Genomic Map