Assay Details
Target Gene Details
Entrez Gene ID: | 51246 |
Gene Name: | shisa family member 5 |
Gene Aliases: |
SCOTIN |
Location: |
Chr.3:48467798-48504826 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHISA5 | NM_001272066.1 | NP_001258995.1 | ||
| NM_001272067.1 | NP_001258996.1 | |||
| XM_005265205.1 | XP_005265262.1 | |||
| AK056328.1 | BAB71152.1 | |||
| BX344125.2 | ||||
| DA020280.1 | ||||
| DA025113.1 | ||||
| DA045437.1 | ||||
| DA657994.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834684 | Chr.3:48344209 - 48551890 on Build GRCh38 | Gain |
|
| nsv523981 | Chr.3:48478880 - 48683607 on Build GRCh38 | Loss |
|
| nsv590242 | Chr.3:48392157 - 48518906 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs146502075] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map