Assay Details
Target Gene Details
Entrez Gene ID: | 80321 |
Gene Name: | centrosomal protein 70 |
Gene Aliases: |
BITE |
Location: |
Chr.3:138494339-138594383 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv999041 | Chr.3:138429970 - 138629146 on Build GRCh38 | Gain |
|
| nsv428422 | Chr.3:137344347 - 138686784 on Build GRCh38 | Gain+Loss |
|
| esv3597916 | Chr.3:138478438 - 138638584 on Build GRCh38 | Gain |
|
| dgv4908n100 | Chr.3:138494324 - 139018846 on Build GRCh38 | Gain |
|
| nsv522517 | Chr.3:138455681 - 138579140 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map