Assay Details
Target Gene Details
Entrez Gene ID: | 100288428 |
Gene Name: | LMCD1 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.3:8221147-8501658 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMCD1-AS1 | NR_033378.1 | |||
| AK124088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3595211 | Chr.3:8222473 - 8225726 on Build GRCh38 | Loss |
|
| nsv1007359 | Chr.3:7726973 - 8302289 on Build GRCh38 | Loss |
|

Genomic Map