Assay Details
Target Gene Details
Entrez Gene ID: | 285386 |
Gene Name: | tumor protein p63 regulated 1 |
Gene Aliases: |
FAM79B |
Location: |
Chr.3:189000774-189325304 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPRG1 | XM_017006257.1 | XP_016861746.1 | ||
| XM_017006258.1 | XP_016861747.1 | |||
| XM_017006261.1 | XP_016861750.1 | |||
| AK123232.1 | BAC85565.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73053445] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map