Assay Details
Target Gene Details
Entrez Gene ID: | 2720 |
Gene Name: | galactosidase beta 1 |
Gene Aliases: |
EBP, ELNR1, MPS4B |
Location: |
Chr.3:32996608-33097230 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GLB1 | NM_000404.3 | NP_000395.2 | ||
NM_001079811.2 | NP_001073279.1 | |||
NM_001135602.2 | NP_001129074.1 | |||
NM_001317040.1 | NP_001303969.1 | |||
AK130133.1 | ||||
AK222763.1 | BAD96483.1 | |||
AK223091.1 | BAD96811.1 | |||
AK298985.1 | ||||
AK300021.1 | ||||
AK300753.1 | ||||
AK312988.1 | ||||
BC007493.2 | AAH07493.1 | |||
BT007147.1 | AAP35811.1 | |||
M22590.1 | AAA51822.1 | |||
M27507.1 | AAA51819.1 | |||
M27508.1 | AAA35599.1 | |||
M34423.1 | AAA51823.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv998302 | Chr.3:32595752 - 33112754 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1003285 | Chr.3:31386933 - 33172886 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv3753 | Chr.3:32966390 - 33027225 on Build GRCh38 | Insertion |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
