Assay Details
Target Gene Details
Entrez Gene ID: | 105377135 |
Gene Name: | transcription initiation factor TFIID subunit 4-like |
Gene Aliases: |
- |
Location: |
Chr.3:52999045-53048252 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105377135 | XM_011534353.2 | XP_011532655.1 |
Target Gene Details
Entrez Gene ID: | 51460 |
Gene Name: | Scm-like with four mbt domains 1 |
Gene Aliases: |
RU1, SFMBT, hSFMBT |
Location: |
Chr.3:52899209-53046655 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SFMBT1 | NM_016329.3 | NP_057413.2 | ||
| XM_005265221.3 | XP_005265278.1 | |||
| XM_006713203.3 | XP_006713266.1 | |||
| XM_006713204.3 | XP_006713267.1 | |||
| XM_011533824.2 | XP_011532126.1 | |||
| XM_011533825.2 | XP_011532127.1 | |||
| AF168132.1 | AAF19794.1 | |||
| AK313965.1 | ||||
| BC014614.2 | AAH14614.1 | |||
| BG717319.1 | ||||
| BG723139.1 | ||||
| BU553665.1 | ||||
| CR981845.1 | ||||
| DB064119.1 | ||||
| DB070525.1 | ||||
| DB081417.1 | ||||
| DB085221.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv437713 | Chr.3:52992698 - 53021506 on Build GRCh38 | Loss |
|
| dgv8331n54 | Chr.3:52993967 - 53028645 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map