Assay Details
Target Gene Details
Entrez Gene ID: | 5978 |
Gene Name: | RE1 silencing transcription factor |
Gene Aliases: |
NRSF, WT6, XBR |
Location: |
Chr.4:56907876-56935847 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REST | NM_005612.4 | NP_005603.3 | ||
| XM_017008527.1 | XP_016864016.1 | |||
| BC038985.1 | AAH38985.1 | |||
| BC132859.1 | ||||
| BC136491.1 | ||||
| JX896957.1 | ||||
| JX896958.1 | ||||
| JX896959.1 | ||||
| JX896960.1 | ||||
| JX896961.1 | ||||
| JX896962.1 | ||||
| JX896963.1 | ||||
| JX896964.1 | ||||
| JX896965.1 | ||||
| JX896971.1 | ||||
| JX896972.1 | ||||
| JX896973.1 | ||||
| JX896974.1 | ||||
| JX896975.1 | ||||
| JX896976.1 | ||||
| JX896977.1 | ||||
| JX896978.1 | ||||
| JX896979.1 | ||||
| JX896980.1 | ||||
| JX896983.1 | ||||
| JX896984.1 | ||||
| JX896985.1 | ||||
| JX896986.1 | ||||
| JX896987.1 | ||||
| JX896988.1 | ||||
| JX896991.1 | ||||
| KC117266.1 | ||||
| U13879.1 | AAC50115.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5214n100 | Chr.4:56786612 - 56999637 on Build GRCh38 | Gain |
|
| nsv594205 | Chr.4:56907276 - 56908898 on Build GRCh38 | Gain |
|
| nsv822562 | Chr.4:56902489 - 56913549 on Build GRCh38 | Gain |
|
| nsv1074717 | Chr.4:56905133 - 56910334 on Build GRCh38 | Deletion |
|
| esv22393 | Chr.4:56906104 - 56909418 on Build GRCh38 | Loss |
|
| nsv594206 | Chr.4:56907429 - 56909377 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map