Assay Details
Target Gene Details
Entrez Gene ID: | 2891 |
Gene Name: | glutamate ionotropic receptor AMPA type subunit 2 |
Gene Aliases: |
GLUR2, GLURB, GluA2, GluR-K2, HBGR2 |
Location: |
Chr.4:157205099-157370583 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIA2 | XM_017008115.1 | XP_016863604.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1018387 | Chr.4:157038779 - 157843038 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map