Assay Details
Target Gene Details
Entrez Gene ID: | 64850 |
Gene Name: | ethanolamine-phosphate phospho-lyase |
Gene Aliases: |
AGXT2L1 |
Location: |
Chr.4:108742040-108763180 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ETNPPL | NM_001146590.1 | NP_001140062.1 | ||
| NM_001146627.1 | NP_001140099.1 | |||
| NM_031279.3 | NP_112569.2 | |||
| NR_027474.1 | ||||
| NR_027475.1 | ||||
| XM_006714290.2 | XP_006714353.1 | |||
| AJ298293.1 | CAC22253.1 | |||
| AK091888.1 | BAC03766.1 | |||
| AK294072.1 | ||||
| AK294235.1 | ||||
| BC022526.1 | AAH22526.1 | |||
| DC311077.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519226 | Chr.4:108739559 - 108766158 on Build GRCh38 | Loss |
|
| nsv1000660 | Chr.4:108730447 - 108787621 on Build GRCh38 | Gain |
|
| nsv830034 | Chr.4:108736535 - 108898569 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map