Assay Details
Target Gene Details
Entrez Gene ID: | 5602 |
Gene Name: | mitogen-activated protein kinase 10 |
Gene Aliases: |
JNK3, JNK3A, PRKM10, SAPK1b, p493F12, p54bSAPK |
Location: |
Chr.4:86012296-86594117 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
More Information
Set Membership: |
|

Genomic Map