Assay Details
Target Gene Details
Entrez Gene ID: | 79931 |
Gene Name: | TNFAIP3 interacting protein 3 |
Gene Aliases: |
ABIN-3, LIND |
Location: |
Chr.4:121127161-121227609 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNIP3 | NM_001128843.2 | NP_001122315.2 | ||
| NM_001244764.1 | NP_001231693.1 | |||
| NM_024873.5 | NP_079149.3 | |||
| XM_011532256.2 | XP_011530558.1 | |||
| XM_011532257.2 | XP_011530559.1 | |||
| XM_017008619.1 | XP_016864108.1 | |||
| XM_017008620.1 | XP_016864109.1 | |||
| XM_017008621.1 | XP_016864110.1 | |||
| XM_017008622.1 | XP_016864111.1 | |||
| XM_017008623.1 | XP_016864112.1 | |||
| XM_017008624.1 | XP_016864113.1 | |||
| XM_017008625.1 | XP_016864114.1 | |||
| AF277289.1 | AAL02151.1 | |||
| AJ320534.1 | CAC85929.1 | |||
| AK290409.1 | ||||
| AK301058.1 | ||||
| AK303192.1 | ||||
| BC128408.1 | ||||
| BP335455.1 | ||||
| DC389224.1 | ||||
| EF585496.1 |
More Information
Set Membership: |
|

Genomic Map