Assay Details
Target Gene Details
Entrez Gene ID: | 6622 |
Gene Name: | synuclein alpha |
Gene Aliases: |
NACP, PARK1, PARK4, PD1 |
Location: |
Chr.4:89724099-89838324 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNCA | NM_000345.3 | NP_000336.1 | ||
| NM_001146054.1 | NP_001139526.1 | |||
| NM_001146055.1 | NP_001139527.1 | |||
| NM_007308.2 | NP_009292.1 | |||
| XM_011532208.2 | XP_011530510.1 | |||
| XM_017008562.1 | XP_016864051.1 | |||
| XM_017008563.1 | XP_016864052.1 | |||
| AK290169.1 | ||||
| AY049786.1 | AAL15443.1 | |||
| BC013293.2 | AAH13293.1 | |||
| BC108275.1 | AAI08276.1 | |||
| CR457058.1 | CAG33339.1 | |||
| CR541653.1 | CAG46454.1 | |||
| D31839.1 | BAA06625.1 | |||
| DA391115.1 | ||||
| JN709859.1 | ||||
| JN709860.1 | ||||
| JN709861.1 | ||||
| JN709862.1 | ||||
| JN709863.1 | ||||
| JN709864.1 | ||||
| JN709865.1 | ||||
| JN709866.1 | ||||
| JN709867.1 | ||||
| JN709868.1 | ||||
| JN709869.1 | ||||
| L08850.1 | AAA16117.1 | |||
| L36674.1 | AAA98493.1 | |||
| L36675.1 | AAA98487.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012406 | Chr.4:89246630 - 90245636 on Build GRCh38 | Loss |
|
| nsv594806 | Chr.4:88621130 - 90539537 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map