Assay Details
Target Gene Details
Entrez Gene ID: | 84740 |
Gene Name: | AFAP1 antisense RNA 1 |
Gene Aliases: |
AFAP1-AS, AFAP1AS |
Location: |
Chr.4:7754090-7778928 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AFAP1-AS1 | NR_026892.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2727075 | Chr.4:7756307 - 7756847 on Build GRCh38 | Deletion |
|
| esv2727074 | Chr.4:7756158 - 7757234 on Build GRCh38 | Deletion |
|

Genomic Map