Assay Details
Target Gene Details
Entrez Gene ID: | 2638 |
Gene Name: | GC, vitamin D binding protein |
Gene Aliases: |
DBP, DBP/GC, GRD3, Gc-MAF, GcMAF, HEL-S-51, VDBG, VDBP |
Location: |
Chr.4:71741693-71805520 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GC | NM_000583.3 | NP_000574.2 | ||
| NM_001204306.1 | NP_001191235.1 | |||
| NM_001204307.1 | NP_001191236.1 | |||
| XM_006714177.2 | XP_006714240.1 | |||
| AK223458.1 | BAD97178.1 | |||
| AK290827.1 | ||||
| AK298433.1 | ||||
| AK309595.1 | ||||
| AK315853.1 | ||||
| BC057228.1 | AAH57228.1 | |||
| EU794674.1 | ||||
| X03178.1 | CAA26938.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829967 | Chr.4:71602549 - 71760380 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map