Assay Details
Target Gene Details
Entrez Gene ID: | 441009 |
Gene Name: | CPEB2 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.4:14909961-15002045 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPEB2-AS1 | NR_038857.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv593756 | Chr.4:14896504 - 14935342 on Build GRCh38 | Loss |
|

Genomic Map