Assay Details
Target Gene Details
Entrez Gene ID: | 55132 |
Gene Name: | La ribonucleoprotein domain family member 1B |
Gene Aliases: |
LARP2 |
Location: |
Chr.4:128060655-128222931 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LARP1B | XM_017008340.1 | 1 | 256 | XP_016863829.1 |
| XM_017008344.1 | 1 | 256 | XP_016863833.1 | |
| XM_017008353.1 | 1 | 256 | XP_016863842.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv24873 | Chr.4:128060903 - 128062238 on Build GRCh38 | Loss |
|
| dgv5385n100 | Chr.4:127882004 - 128173863 on Build GRCh38 | Gain |
|
| nsv595388 | Chr.4:127932758 - 128163676 on Build GRCh38 | Gain |
|
| dgv9258n54 | Chr.4:128060930 - 128061604 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map