Assay Details
Target Gene Details
Entrez Gene ID: | 100507266 |
Gene Name: | STX18 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.4:4542131-4710938 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STX18-AS1 | NR_037888.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1007737 | Chr.4:4513829 - 4546750 on Build GRCh38 | Gain |
|
| nsv521279 | Chr.4:4532247 - 4633549 on Build GRCh38 | Loss |
|
| esv3599463 | Chr.4:4543982 - 4553020 on Build GRCh38 | Loss |
|

Genomic Map